Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969086.1 | XP_027824887.1 | 2 - 3 | 230 | 101 | |
| XM_027969087.1 | XP_027824888.1 | 4 - 5 | 621 | 101 | ||
| XM_027969088.1 | XP_027824889.1 | 4 - 5 | 600 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027969086.1 transcripts.
Set Membership: |
|

DYNC2I1
Genomic Map