Assay Details
| Gene Symbol: SLC16A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012185605.3 | XP_012040995.3 | 4 - 5 | 1322 | 98 | |
| XM_027975010.1 | XP_027830811.1 | 4 - 5 | 1316 | 98 | ||
| XM_027975011.1 | XP_027830812.1 | 4 - 5 | 1976 | 98 | ||
More Information
Set Membership: |
|

SLC16A11
Genomic Map