Assay Details
| Gene Symbol: AMZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973985.1 | XP_027829786.1 | 6 - 7 | 1321 | 61 | |
| XM_027973986.1 | XP_027829787.1 | 7 - 8 | 1036 | 61 | ||
More Information
Set Membership: |
|

AMZ2
Genomic Map