Assay Details
| Gene Symbol: F9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022305.4 | XP_004022354.1 | 6 - 7 | 753 | 68 | |
| GenBank mRNA | M26233.1 | - | 1 - 2 | 189 | 68 | |
More Information
Set Membership: |
|

F9
Genomic Map