Assay Details
| Gene Symbol: LOC101104231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008016.4 | XP_004008065.1 | 17 - 18 | 2843 | 103 | |
| XM_004008018.4 | XP_004008067.1 | 18 - 19 | 2864 | 103 | ||
| XM_004008019.4 | XP_004008068.1 | 19 - 20 | 2874 | 103 | ||
| XM_012177040.3 | XP_012032430.1 | 19 - 20 | 2874 | 103 | ||
| XM_012177053.3 | XP_012032443.1 | 19 - 20 | 2873 | 103 | ||
| XM_012177054.3 | XP_012032444.1 | 17 - 18 | 2843 | 103 | ||
| XM_027968768.1 | XP_027824569.1 | 18 - 19 | 2852 | 103 | ||
More Information
Set Membership: |
|

LOC101104231
Genomic Map