Assay Details
| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101694.2 | XP_014957180.1 | 4 - 5 | 353 | 79 | |
| XM_027980286.1 | XP_027836087.1 | 3 - 4 | 361 | 79 | ||
More Information
Set Membership: |
|

CRYBB2
Genomic Map