Assay Details
| Gene Symbol: LOC101103973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177031.3 | XP_012032421.2 | 18 - 19 | 2124 | 77 | |
| XM_027968763.1 | XP_027824564.1 | 17 - 18 | 4723 | 77 | ||
| XM_027968764.1 | XP_027824565.1 | 15 - 16 | 1781 | 77 | ||
| XM_027968765.1 | XP_027824566.1 | 16 - 17 | 1710 | 77 | ||
More Information
Set Membership: |
|

LOC101103973
Genomic Map