Assay Details
| Gene Symbol: NDUFAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011828.3 | XP_004011877.2 | 1 - 2 | 208 | 85 | |
| XM_027973208.1 | XP_027829009.1 | 1 - 2 | 190 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027973208.1 transcripts.
Set Membership: |
|

NDUFAF6
Genomic Map