Assay Details
| Gene Symbol: JOSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977534.1 | XP_027833335.1 | 3 - 4 | 439 | 72 | |
| XM_027977535.1 | XP_027833336.1 | 3 - 4 | 495 | 72 | ||
More Information
Set Membership: |
|

JOSD2
Genomic Map