Assay Details
| Gene Symbol: EFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017779.3 | XP_004017828.1 | 3 - 4 | 284 | 63 | |
| XM_012098550.3 | XP_011953940.1 | 3 - 4 | 260 | 63 | ||
More Information
Set Membership: |
|

EFL1
Genomic Map