Assay Details
| Gene Symbol: CALHM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020164.4 | XP_004020213.2 | 2 - 3 | 548 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CALHM3| Gene Symbol: CALHM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020164.4 | XP_004020213.2 | 2 - 3 | 548 | 70 | |
Set Membership: |
|