Assay Details
| Gene Symbol: CCDC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974305.1 | XP_027830106.1 | 18 - 19 | 2854 | 58 | |
| XM_027974306.1 | XP_027830107.1 | 17 - 18 | 2742 | 58 | ||
| XM_027974307.1 | XP_027830108.1 | 16 - 17 | 2633 | 58 | ||
More Information
Set Membership: |
|

CCDC57
Genomic Map