Assay Details
| Gene Symbol: UCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012185487.3 | XP_012040877.2 | 6 - 7 | 629 | 65 | |
| XM_027977701.1 | XP_027833502.1 | 6 - 7 | 902 | 65 | ||
More Information
Set Membership: |
|

UCK2
Genomic Map