Assay Details
| Gene Symbol: WDR54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007319.4 | XP_004007368.4 | 4 - 5 | 497 | 66 | |
| XM_027967022.1 | XP_027822823.1 | 4 - 5 | 497 | 66 | ||
| XM_027967023.1 | XP_027822824.1 | 3 - 4 | 434 | 66 | ||
More Information
Set Membership: |
|

WDR54
Genomic Map