Assay Details
| Gene Symbol: MAPRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020434.4 | XP_004020483.1 | 4 - 5 | 779 | 63 | |
| XM_004020435.4 | XP_004020484.2 | 5 - 6 | 693 | 63 | ||
| XM_012103351.3 | XP_011958741.1 | 6 - 7 | 683 | 63 | ||
| XM_012103352.3 | XP_011958742.1 | 5 - 6 | 820 | 63 | ||
More Information
Set Membership: |
|

MAPRE2
Genomic Map