Assay Details
Gene Symbol: LOC101122314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958416.1 | XP_027814217.1 | 1 | 1073 | 101 |
Gene Symbol: ENDOU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012174391.3 | XP_012029781.2 | 9 - 10 | 101 | ||
XM_012174392.3 | XP_012029782.2 | 8 - 9 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
