Assay Details
| Gene Symbol: LOC101112585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013721.4 | XP_004013770.1 | 5 - 6 | 969 | 67 | |
| XM_012187328.2 | XP_012042718.1 | 4 - 5 | 592 | 67 | ||
| XM_012187331.2 | XP_012042721.1 | 4 - 5 | 531 | 67 | ||
| XM_012187332.2 | XP_012042722.1 | 4 - 5 | 550 | 67 | ||
| XM_027975640.1 | XP_027831441.1 | 4 - 5 | 1033 | 67 | ||
More Information
Set Membership: |
|

LOC101112585
Genomic Map