Assay Details
Gene Symbol: HS1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966541.1 | XP_027822342.1 | 4 - 5 | 671 | 90 | |
XM_027966542.1 | XP_027822343.1 | 4 - 5 | 677 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
