Assay Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178675.3 | XP_012034065.1 | 2 - 3 | 1621 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCTD16| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178675.3 | XP_012034065.1 | 2 - 3 | 1621 | 122 | |
Set Membership: |
|