Assay Details
| Gene Symbol: LACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012184387.2 | XP_012039777.2 | 2 - 3 | 809 | 84 | |
| XM_027973535.1 | XP_027829336.1 | 2 - 3 | 686 | 84 | ||
More Information
Set Membership: |
|

LACC1
Genomic Map