Assay Details
| Gene Symbol: PLPPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969684.1 | XP_027825485.1 | 7 - 8 | 1030 | 61 | |
| XM_027969686.1 | XP_027825487.1 | 9 - 10 | 1348 | 61 | ||
| XM_027969687.1 | XP_027825488.1 | 6 - 7 | 840 | 61 | ||
| XM_027969688.1 | XP_027825489.1 | 7 - 8 | 1014 | 61 | ||
| XM_027969689.1 | XP_027825490.1 | 9 - 10 | 1326 | 61 | ||
| XM_027969690.1 | XP_027825491.1 | 9 - 10 | 1409 | 61 | ||
More Information
Set Membership: |
|

PLPPR2
Genomic Map