Assay Details
| Gene Symbol: RNF168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003008.3 | XP_004003057.1 | 6 - 7 | 1129 | 71 | |
| XM_027956964.1 | XP_027812765.1 | 6 - 7 | 1128 | 71 | ||
More Information
Set Membership: |
|

RNF168
Genomic Map