Assay Details
Gene Symbol: LOC101119820 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009624.4 | XP_004009673.2 | 4 - 5 | 382 | 100 | |
XM_012179515.3 | XP_012034905.2 | 4 - 5 | 355 | 100 |
Gene Symbol: CASP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970817.1 | XP_027826618.1 | 4 - 5 | 362 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
