Assay Details
| Gene Symbol: RIPOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977315.1 | XP_027833116.1 | 16 - 17 | 2095 | 61 | |
| XM_027977316.1 | XP_027833117.1 | 15 - 16 | 2195 | 61 | ||
| XM_027977317.1 | XP_027833118.1 | 15 - 16 | 2181 | 61 | ||
| XM_027977318.1 | XP_027833119.1 | 14 - 15 | 2060 | 61 | ||
More Information
Set Membership: |
|

RIPOR3
Genomic Map