Assay Details
| Gene Symbol: FKBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004544.4 | XP_004004593.2 | 1 - 2 | 327 | 75 | |
| XM_012132608.3 | XP_011987998.2 | 1 - 2 | 333 | 75 | ||
More Information
Set Membership: |
|

FKBP7
Genomic Map