Assay Details
| Gene Symbol: TFPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005355.4 | XP_004005404.3 | 2 - 3 | 359 | 77 | |
| XM_027964720.1 | XP_027820521.1 | 2 - 3 | 361 | 77 | ||
More Information
Set Membership: |
|

TFPI
Genomic Map