Assay Details
| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978472.1 | XP_027834273.1 | 3 - 4 | 290 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAPPC6A| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978472.1 | XP_027834273.1 | 3 - 4 | 290 | 77 | |
Set Membership: |
|