Assay Details
| Gene Symbol: ZNF268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097619.2 | XP_011953009.2 | 5 - 6 | 629 | 65 | |
| XM_027956350.1 | XP_027812151.1 | 4 - 5 | 913 | 65 | ||
More Information
Set Membership: |
|

ZNF268
Genomic Map