Assay Details
| Gene Symbol: PELI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959625.1 | XP_027815426.1 | 5 - 6 | 586 | 86 | |
| XM_027959626.1 | XP_027815427.1 | 4 - 5 | 573 | 86 | ||
| XM_027959627.1 | XP_027815428.1 | 4 - 5 | 497 | 86 | ||
More Information
Set Membership: |
|

PELI3
Genomic Map