Assay Details
| Gene Symbol: LOC101117299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961207.1 | XP_027817008.1 | 3 - 4 | 525 | 83 | |
| XM_027961208.1 | XP_027817009.1 | 3 - 4 | 336 | 83 | ||
More Information
Set Membership: |
|

LOC101117299
Genomic Map