Assay Details
| Gene Symbol: LOC101117299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961207.1 | XP_027817008.1 | 12 - 13 | 1471 | 66 | |
| XM_027961208.1 | XP_027817009.1 | 12 - 13 | 1282 | 66 | ||
More Information
Set Membership: |
|

LOC101117299
Genomic Map