Assay Details
| Gene Symbol: CRNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975162.1 | XP_027830963.1 | 2 - 3 | 239 | 63 | |
| XM_027975169.1 | XP_027830970.1 | 3 - 4 | 255 | 63 | ||
| XM_027975174.1 | XP_027830975.1 | 2 - 3 | 222 | 63 | ||
| XM_027975175.1 | XP_027830976.1 | 3 - 4 | 231 | 63 | ||
| XM_027975179.1 | XP_027830980.1 | 2 - 3 | 212 | 63 | ||
| XM_027975184.1 | XP_027830985.1 | 3 - 4 | 227 | 63 | ||
| XM_027975191.1 | XP_027830992.1 | 2 - 3 | 239 | 63 | ||
| XM_027975202.1 | XP_027831003.1 | 2 - 3 | 239 | 63 | ||
More Information
Set Membership: |
|

CRNN
Genomic Map