Assay Details
| Gene Symbol: LOC101103413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014383.3 | XP_004014432.1 | 13 - 14 | 2096 | 95 | |
| XM_027976880.1 | XP_027832681.1 | 12 - 13 | 2183 | 95 | ||
| Gene Symbol: LOC114117593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976877.1 | XP_027832678.1 | 13 - 14 | 2054 | 95 | |
More Information
Set Membership: |
|

LOC101103413
Genomic Map