Assay Details
| Gene Symbol: KCNMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016863.4 | XP_004016912.1 | 3 - 4 | 445 | 109 | |
| XM_027979977.1 | XP_027835778.1 | 2 - 3 | 570 | 109 | ||
More Information
Set Membership: |
|

KCNMB1
Genomic Map