Assay Details
| Gene Symbol: CXHXorf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962952.1 | XP_027818753.1 | 6 - 7 | 1013 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXHXorf38| Gene Symbol: CXHXorf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962952.1 | XP_027818753.1 | 6 - 7 | 1013 | 106 | |
Set Membership: |
|