Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958350.1 | XP_027814151.1 | 7 - 8 | 1609 | 70 | |
| XM_027958351.1 | XP_027814152.1 | 6 - 7 | 1480 | 70 | ||
More Information
Set Membership: |
|

RNF8
Genomic Map