Assay Details
| Gene Symbol: POU4F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098229.2 | XP_014953715.2 | 1 - 2 | 139 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POU4F1| Gene Symbol: POU4F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098229.2 | XP_014953715.2 | 1 - 2 | 139 | 101 | |
Set Membership: |
|