Assay Details
| Gene Symbol: ZC3H12D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972541.1 | XP_027828342.1 | 5 - 6 | 1103 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZC3H12D| Gene Symbol: ZC3H12D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972541.1 | XP_027828342.1 | 5 - 6 | 1103 | 95 | |
Set Membership: |
|