Assay Details
| Gene Symbol: CDC42SE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015095934.2 | XP_014951420.1 | 5 - 6 | 801 | 71 | |
| XM_015095935.2 | XP_014951421.1 | 6 - 7 | 858 | 71 | ||
| XM_015095936.2 | XP_014951422.1 | 4 - 5 | 619 | 71 | ||
| XM_027969959.1 | XP_027825760.1 | 5 - 6 | 711 | 71 | ||
More Information
Set Membership: |
|

CDC42SE2
Genomic Map