Assay Details
| Gene Symbol: MIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078655.1 | NP_001072123.1 | 1 - 2 | 110 | 94 | |
| GenBank mRNA | DQ414692.1 | - | 1 - 2 | 110 | 94 | |
More Information
Set Membership: |
|

MIF
Genomic Map