Assay Details
| Gene Symbol: FMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187270.3 | XP_012042660.2 | 5 - 6 | 1110 | 69 | |
| XM_027975597.1 | XP_027831398.1 | 5 - 6 | 1115 | 69 | ||
More Information
Set Membership: |
|

FMO4
Genomic Map