Assay Details
| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019677.4 | XP_004019726.2 | 1 - 2 | 265 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNASEH2C| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019677.4 | XP_004019726.2 | 1 - 2 | 265 | 105 | |
Set Membership: |
|