Assay Details
| Gene Symbol: UNC5CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958931.1 | XP_027814732.1 | 5 - 6 | 1333 | 59 | |
| XM_027958932.1 | XP_027814733.1 | 4 - 5 | 1473 | 59 | ||
More Information
Set Membership: |
|

UNC5CL
Genomic Map