Assay Details
| Gene Symbol: WIPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098766.2 | XP_014954252.1 | 4 - 5 | 557 | 59 | |
| XM_015098767.2 | XP_014954253.1 | 4 - 5 | 520 | 59 | ||
| XM_015098770.2 | XP_014954256.1 | 3 - 4 | 396 | 59 | ||
| XM_015098773.2 | XP_014954259.2 | 4 - 5 | 557 | 59 | ||
| XM_027974582.1 | XP_027830383.1 | 3 - 4 | 433 | 59 | ||
More Information
Set Membership: |
|

WIPF2
Genomic Map