Assay Details
| Gene Symbol: HMGCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002390.4 | XP_004002439.1 | 6 - 7 | 1255 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HMGCS2| Gene Symbol: HMGCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002390.4 | XP_004002439.1 | 6 - 7 | 1255 | 61 | |
Set Membership: |
|