Assay Details
| Gene Symbol: SLC36A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970431.1 | XP_027826232.1 | 4 - 5 | 452 | 77 | |
| XM_027970432.1 | XP_027826233.1 | 4 - 5 | 452 | 77 | ||
| XM_027970433.1 | XP_027826234.1 | 4 - 5 | 452 | 77 | ||
| XM_027970434.1 | XP_027826235.1 | 4 - 5 | 233 | 77 | ||
More Information
Set Membership: |
|

SLC36A1
Genomic Map