Assay Details
| Gene Symbol: FAM3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012099781.3 | XP_011955171.1 | 4 - 5 | 329 | 67 | |
| XM_012099782.3 | XP_011955172.1 | 4 - 5 | 359 | 67 | ||
| XM_012099784.2 | XP_011955174.1 | 4 - 5 | 329 | 67 | ||
More Information
Set Membership: |
|

FAM3D
Genomic Map