Assay Details
| Gene Symbol: ZFP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959134.1 | XP_027814935.1 | 3 - 4 | 1639 | 67 | |
| XM_027959135.1 | XP_027814936.1 | 4 - 5 | 842 | 67 | ||
| XM_027959136.1 | XP_027814937.1 | 5 - 6 | 799 | 67 | ||
| XM_027959137.1 | XP_027814938.1 | 4 - 5 | 752 | 67 | ||
| XM_027959138.1 | XP_027814939.1 | 4 - 5 | 1071 | 67 | ||
More Information
Set Membership: |
|

ZFP57
Genomic Map