Assay Details
| Gene Symbol: RELCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960868.1 | XP_027816669.1 | 22 - 23 | 3051 | 66 | |
| XM_027960869.1 | XP_027816670.1 | 21 - 22 | 3027 | 66 | ||
| XM_027960870.1 | XP_027816671.1 | 21 - 22 | 3027 | 66 | ||
| XM_027960871.1 | XP_027816672.1 | 21 - 22 | 2979 | 66 | ||
| XM_027960872.1 | XP_027816673.1 | 20 - 21 | 2955 | 66 | ||
| XM_027960873.1 | XP_027816674.1 | 22 - 23 | 3051 | 66 | ||
| XM_027960874.1 | XP_027816675.1 | 22 - 23 | 3051 | 66 | ||
More Information
Set Membership: |
|

RELCH
Genomic Map