Assay Details
| Gene Symbol: ASIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006367.4 | XP_004006416.1 | 9 - 10 | 1372 | 66 | |
| XM_004006368.4 | XP_004006417.1 | 8 - 9 | 1514 | 66 | ||
More Information
Set Membership: |
|

ASIC1
Genomic Map